Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Carbonic Anhydrase II Polyclonal Antibody
Sheep Polyclonal Antibody
Supplier: Invitrogen™ PA533167
Description
In Western blot, this antibody detects a band of ~18 kDa in rat adrenomedullary chromaffin cell lysate.
CA2 is one of several (at least 7) isozymes of carbonic anhydrase. Carbonic anhydrase catalyzes reversible hydration of carbon dioxide. Defects in this enzyme are associated with osteopetrosis and renal tubular acidosis.
Specifications
Carbonic Anhydrase II | |
Polyclonal | |
glycine buffered saline with 0.01% benzamidine, 0.1% EACA, 1mM EDTA and <0.1% sodium azide | |
P00918, P00920 | |
CA2, Car2 | |
Purified human carbonic anhydrase II (CAII) prepared from erythrocytes. | |
1 mL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Immunodiffusion, Western Blot, Immunocytochemistry | |
Unconjugated | |
CA2 | |
AI131712; CA II; CA2; CAC; CAII; CA-II; Car2; Car-2; Carbonate dehydratase II; carbonic anhydrase 2; carbonic anhydrase B; carbonic anhydrase C; Carbonic anhydrase II; carbonic dehydratase; epididymis luminal protein 76; epididymis secretory protein Li 282; HEL-76; HEL-S-282; Ltw-5; Lvtw-5 | |
Sheep | |
Ion-exchange chromatography | |
RUO | |
12349, 760 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction