Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Carbonic Anhydrase II Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551598
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551598 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551598 Supplier Invitrogen™ Supplier No. PA551598
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YGKHNGPEHW HKDFPIAKGE RQSPVDIDTH TAKYDPSLKP LSVSYDQATS LRILNNGHAF NVEFDDSQDK AVLKGGPLDG TYRLIQFHFH WGSLDGQGSE HTVDKKKYAA ELHLVHWNTK YGDFGKAVQQ PDGLAVLG Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 80%.

CA2 is one of several (at least 7) isozymes of carbonic anhydrase. Carbonic anhydrase catalyzes reversible hydration of carbon dioxide. Defects in this enzyme are associated with osteopetrosis and renal tubular acidosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Carbonic Anhydrase II
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CA2
Gene Accession No. P00918, P00920, P27139
Gene Alias AI131712; CA II; CA2; CAC; CAII; CA-II; Car2; Car-2; Carbonate dehydratase II; carbonic anhydrase 2; carbonic anhydrase B; carbonic anhydrase C; Carbonic anhydrase II; carbonic dehydratase; epididymis luminal protein 76; epididymis secretory protein Li 282; HEL-76; HEL-S-282; Ltw-5; Lvtw-5
Gene Symbols CA2, Car2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Carbonic Anhydrase II. Recombinant protein control fragment (Product #RP-95151).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12349, 54231, 760
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.