Learn More
Invitrogen™ Carbonic Anhydrase II Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA589803
Description
Immunogen sequence: MSHHWGYGKH NGPEHWHKDF PIAKGERQSP VDIDTHTAKY DPSLKPLSVS YDQATSLRIL NNGHAFNVEF DDSQDKAVLK GGPLDGTYRL IQFHFHWGSL DGQGSEHTVD KKKYAAELHL VHWNTKYGDF GKAVQQPDGL AVLGIFLKVG SAKPGLQKVV DVLDSIKTKG KSADFTNFDP RGLLPESLDY WTYPGSLTTP PLLECVTWIV LKEPISVSSE QVLKFRKLNF NGEGEPEELM VDNWRPAQPL; Positive Samples: HL-60, 293T, Mouse kidney, Mouse spleen, Mouse lung; Cellular Location: Cell membrane, Cytoplasm.
CA2 is one of several (at least 7) isozymes of carbonic anhydrase. Carbonic anhydrase catalyzes reversible hydration of carbon dioxide. Defects in this enzyme are associated with osteopetrosis and renal tubular acidosis.
Specifications
Carbonic Anhydrase II | |
Polyclonal | |
Unconjugated | |
CA2 | |
AI131712; CA II; CA2; CAC; CAII; CA-II; Car2; Car-2; Carbonate dehydratase II; carbonic anhydrase 2; carbonic anhydrase B; carbonic anhydrase C; Carbonic anhydrase II; carbonic dehydratase; epididymis luminal protein 76; epididymis secretory protein Li 282; HEL-76; HEL-S-282; Ltw-5; Lvtw-5 | |
Rabbit | |
Affinity chromatography | |
RUO | |
12349, 760 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
2.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P00918, P00920 | |
CA2, Car2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-250 of human Carbonic Anhydrase 2 (CA2) (NP_0000581). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.