Learn More
Carbonic Anhydrase II Polyclonal Antibody, HRP, Rockland™

Rabbit Polyclonal Antibody
$178.35 - $626.07
Specifications
| Antigen | Carbonic Anhydrase II |
|---|---|
| Concentration | 1 mg/mL |
| Applications | ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | HRP |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-0956
![]()
|
Rockland Immunochemicals
2004031360100 |
100 μg |
Each for $626.07
|
|
|||||
50-199-0957
![]()
|
Rockland Immunochemicals
200403136S |
25 μL |
Each for $178.35
|
|
|||||
Description
Store vial at 4°C prior to restoration. For extended storage aliquot contents and freeze at -20°C or below. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Carbonic anhydrase II is an IgG fraction antibody purified from monospecific antiserum by a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer stated above. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Peroxidase, anti-Rabbit Serum as well as purified and partially purified Carbonic Anhydrase II [Human Erythrocytes]. Cross reactivity against Carbonic Anhydrase II from other sources may occur but have not been specifically determined.
CA2 is one of several (at least 7) isozymes of carbonic anhydrase. Carbonic anhydrase catalyzes reversible hydration of carbon dioxide. Defects in this enzyme are associated with osteopetrosis and renal tubular acidosis.Specifications
| Carbonic Anhydrase II | |
| ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot | |
| HRP | |
| Rabbit | |
| Human | |
| P00918 | |
| 760 | |
| Carbonic Anhydrase II [Human Erythrocytes] | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| CA2 |
| 1 mg/mL | |
| Polyclonal | |
| Lyophilized | |
| RUO | |
| 0.02M potassium phosphate with 0.15M NaCl, 10 mg/mL BSA and 0.01% gentamicin sulfate; pH 7.2 | |
| AI131712; CA II; Ca2; CAC; CAII; CA-II; Car2; Car-2; Carbonate dehydratase II; Carbonic anhydrase 2; carbonic anhydrase B; Carbonic anhydrase C; Carbonic anhydrase II; carbonic dehydratase; epididymis luminal protein 76; epididymis secretory protein Li 282; HEL-76; HEL-S-282; Ltw-5; Lvtw-5 | |
| CA2 | |
| Primary | |
| Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.