Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carbonic Anhydrase III/CA3 Rabbit anti-Human, Clone: 6, Novus Biologicals™

Rabbit Monoclonal Antibody
$353.50 - $560.50
Specifications
Antigen | Carbonic Anhydrase III/CA3 |
---|---|
Clone | 6 |
Dilution | Western Blot 1:500-1:2000, Immunoprecipitation 1-4 ul/mg of lysate |
Applications | Western Blot, Immunoprecipitation |
Classification | Monoclonal |
Description
Carbonic Anhydrase III/CA3 Monoclonal specifically detects Carbonic Anhydrase III/CA3 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
Carbonic Anhydrase III/CA3 | |
Western Blot 1:500-1:2000, Immunoprecipitation 1-4 ul/mg of lysate | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CAIII, CA-III, Car3, Carbonate dehydratase III, carbonic anhydrase 3, Carbonic anhydrase III, carbonic anhydrase III, muscle specific, EC 4.2.1.1 | |
Recombinant Human Carbonic Anhydrase III/CA3 protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
6 | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Lipid and Metabolism | |
0.2 um filtered solution in PBS | |
761 | |
IgG | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title