Learn More
Invitrogen™ Carbonic Anhydrase III Monoclonal Antibody (C2), Invitrogen™

Description
Specifications
Specifications
| Antigen | Carbonic Anhydrase III |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | C2 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 |
| Gene | Ca3 |
| Gene Accession No. | P07451, P14141, P16015, Q5S1S4 |
| Gene Alias | BB219044; CA III; Ca3; CAH3; CAIII; CA-III; CAIII (Muscle); Car3; Car-3; Carbonate dehydratase III; carbonic anhydrase 3; Carbonic anhydrase III; carbonic anhydrase III, muscle specific; carbonic anhydrase-like protein; EC 4.2.1.1; epididymis secretory sperm binding protein Li 167mP; HEL-S-167mP |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.