Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Carbonic Anhydrase III Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101491
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101491 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101491 Supplier Invitrogen™ Supplier No. PA5101491
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CA3.

Carbonic anhydrase III (CAIII) is a member of a multigene family (at least six separate genes are known) that encodes carbonic anhydrase isozymes. These carbonic anhydrases are a class of metalloenzymes that catalyze the reversible hydration of carbon dioxide and are differentially expressed in a number of cell types. The expression of the CA3 gene is strictly tissue specific and present at high levels in skeletal muscle and much lower levels in cardiac and smooth muscle. A proportion of carriers of Duchenne muscle dystrophy have a higher CA3 level than normal. The gene spans 10. 3 kb and contains seven exons and six introns.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Carbonic Anhydrase III
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Ca3
Gene Accession No. P07451, P14141, P16015
Gene Alias BB219044; CA III; Ca3; CAH3; CAIII; CA-III; CAIII (Muscle); Car3; Car-3; Carbonate dehydratase III; carbonic anhydrase 3; Carbonic anhydrase III; carbonic anhydrase III, muscle specific; carbonic anhydrase-like protein; EC 4.2.1.1; epididymis secretory sperm binding protein Li 167mP; HEL-S-167mP
Gene Symbols Ca3, Car3
Host Species Rabbit
Immunogen A synthesized peptide derived from human CA3(Accession P07451), corresponding to amino acid residues G12-N62.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12350, 54232, 761
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.