Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Carbonic Anhydrase III Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA525977

Catalog No. PIPA525977


Only null left
Add to Cart

Description

Description

Carbonic Anhydrase III Polyclonal Antibody for Western Blot, IHC (P)

Carbonic anhydrase III (CAIII) is a member of a multigene family (at least six separate genes are known) that encodes carbonic anhydrase isozymes. These carbonic anhydrases are a class of metalloenzymes that catalyze the reversible hydration of carbon dioxide and are differentially expressed in a number of cell types. The expression of the CA3 gene is strictly tissue specific and present at high levels in skeletal muscle and much lower levels in cardiac and smooth muscle. A proportion of carriers of Duchenne muscle dystrophy have a higher CA3 level than normal. The gene spans 10. 3 kb and contains seven exons and six introns.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Carbonic Anhydrase III
Polyclonal
Unconjugated
Ca3
BB219044; CA III; Ca3; CAH3; CAIII; CA-III; CAIII (Muscle); Car3; Car-3; Carbonate dehydratase III; carbonic anhydrase 3; Carbonic anhydrase III; carbonic anhydrase III, muscle specific; carbonic anhydrase-like protein; EC 4.2.1.1; epididymis secretory sperm binding protein Li 167mP; HEL-S-167mP
Rabbit
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
12350, 761
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
2 mg/mL
PBS with 0.09% sodium azide; pH 7.4
P07451, P16015
Ca3, Car3
KLH conjugated synthetic peptide between 146-177 amino acids from the central region of human CA3.
400 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.