Learn More
Carbonic Anhydrase IIRabbit anti-Bovine, Polyclonal, Rockland™

Rabbit Polyclonal Antibody
Supplier: Rockland Immunochemicals 1004157
Description
Store vial at 4°C prior to restoration. For extended storage aliquot contents and freeze at -20°C or below. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-rabbit serum, purified and partially purified Carbonic Anhydrase II [Bovine Erythrocytes]. Cross reactivity against Carbonic Anhydrase II from other tissues and species may occur but have not been specifically determined.
CA2 is one of several (at least 7) isozymes of carbonic anhydrase. Carbonic anhydrase catalyzes reversible hydration of carbon dioxide. Defects in this enzyme are associated with osteopetrosis and renal tubular acidosis.Specifications
| Carbonic Anhydrase II | |
| Polyclonal | |
| Unconjugated | |
| CA2 | |
| AI131712; CA II; Ca2; CAC; CAII; CA-II; Car2; Car-2; Carbonate dehydratase II; Carbonic anhydrase 2; carbonic anhydrase B; Carbonic anhydrase C; Carbonic anhydrase II; carbonic dehydratase; epididymis luminal protein 76; epididymis secretory protein Li 282; HEL-76; HEL-S-282; Ltw-5; Lvtw-5 | |
| Rabbit | |
| 2 mL | |
| Primary | |
| Bovine | |
| Antibody |
| ELISA, Immunoprecipitation, Western Blot | |
| 85 mg/mL | |
| 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2 | |
| P00921 | |
| CA2 | |
| Carbonic Anhydrase II [Bovine Erythrocytes] | |
| RUO | |
| 280740 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.