Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carbonic Anhydrase IV/CA4 Rabbit anti-Human, Clone: 39, Novus Biologicals™

Rabbit Monoclonal Antibody
$353.50 - $560.50
Specifications
Antigen | Carbonic Anhydrase IV/CA4 |
---|---|
Clone | 39 |
Dilution | ELISA 1:5000-1:10000, Immunohistochemistry, Immunohistochemistry-Paraffin 1:100-1:500 |
Applications | ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Description
Carbonic Anhydrase IV/CA4 Monoclonal specifically detects Carbonic Anhydrase IV/CA4 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Carbonic Anhydrase IV/CA4 | |
ELISA 1:5000-1:10000, Immunohistochemistry, Immunohistochemistry-Paraffin 1:100-1:500 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CAIV, CA-IV, Car4, Carbonate dehydratase IV, carbonic anhydrase 4, carbonic anhydrase IVRP17, carbonic dehydratase IV, EC 4.2.1.1, retinitis pigmentosa 17 (autosomal dominant) | |
Recombinant Human Carbonic Anhydrase IV/CA4 protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
39 | |
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Lipid and Metabolism, Vision | |
0.2 um filtered solution in PBS | |
762 | |
IgG | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title