Learn More
Invitrogen™ Carbonic Anhydrase IV Monoclonal Antibody (310413)

Description
In direct ELISAs and Western blots, less than 10% cross-reactivity with recombinant human (rh) CA1, rhCA2, and recombinant mouse CA4 is observed. This antibody does not cross-react with rhCA3, 8, 9, 10, 12, 13, or 14. Reconstitute at 0.5 mg/mL in sterile PBS.
Specifications
Specifications
| Antigen | Carbonic Anhydrase IV |
| Applications | Immunohistochemistry, Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | 310413 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | CA4 |
| Gene Accession No. | P22748 |
| Gene Alias | AW456718; CA IV; CA4; CAIV; CA-IV; Car4; Carbonate dehydratase IV; carbonic anhydrase 4; Carbonic anhydrase IV; carbonic dehydratase IV; RP17 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.