Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Carbonic Anhydrase IV Monoclonal Antibody (7-2-3#)
Mouse Monoclonal Antibody
$243.00 - $588.50
Specifications
Antigen | Carbonic Anhydrase IV |
---|---|
Clone | 7-2-3# |
Concentration | 1 mg/mL |
Content And Storage | -20°C |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA541560
|
Invitrogen™
MA541560 |
200 μL |
Each for $588.50
|
|
|||||
PIMA543926
|
Invitrogen™
MA543926 |
20 μL |
Each for $243.00
|
|
|||||
Description
Carbonic Anhydrase IV Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
Carbonic anhydrases are a large family of zinc metalloenzymes that catalyze the reversible hydration of carbon dioxide. They participate in a variety of biological processes, including respiration, calcification, acid-base balance, bone resorption, and the formation of aqueous humor, cerebrospinal fluid, saliva, and gastric acid. They show extensive diversity in tissue distribution and in their subcellular localization. This gene encodes a glycosylphosphatidyl-inositol-anchored membrane isozyme expressed on the luminal surfaces of pulmonary capillaries and proximal renal tubules. Its exact function is not known, however, it may have a role in inherited renal abnormalities of bicarbonate transport.Specifications
Carbonic Anhydrase IV | |
1 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
Unconjugated | |
Mouse | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
P48284 | |
29242 | |
Recombinant protein Carbonic Anhydrase IV. The antigen corresponds to amino acid range 20-220 of the target protein. | |
Antibody |
7-2-3# | |
-20°C | |
Monoclonal | |
Liquid | |
IgG2b κ | |
Rat | |
CA4 | |
AW456718; CA IV; CA4; CAIV; CA-IV; Car4; Carbonate dehydratase IV; carbonic anhydrase 4; Carbonic anhydrase IV; carbonic dehydratase IV; RP17 | |
CA4 | |
Primary | |
Protein A/G |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title