Learn More
Invitrogen™ Carbonic Anhydrase IV Polyclonal Antibody, Invitrogen™

Description
Specifications
Specifications
| Antigen | Carbonic Anhydrase IV |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 |
| Gene | CA4 |
| Gene Accession No. | P22748, P48284, Q64444 |
| Gene Alias | AW456718; CA IV; CA4; CAIV; CA-IV; Car4; Carbonate dehydratase IV; carbonic anhydrase 4; Carbonic anhydrase IV; carbonic dehydratase IV; RP17 |
| Gene Symbols | CA4, Car4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.