Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carbonic Anhydrase IX/CA9 Antibody (3), DyLight 755

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289271IR
Description
Carbonic Anhydrase IX/CA9 Monoclonal specifically detects Carbonic Anhydrase IX/CA9 in Human samples. It is validated for Western Blot, ELISA.Specifications
Carbonic Anhydrase IX/CA9 | |
Monoclonal | |
DyLight 755 | |
CAIX, CA-IX, Carbonate dehydratase IX, carbonic anhydrase 9, carbonic anhydrase IX, carbonic dehydratase, EC 4.2.1.1, G250, Membrane antigen MN, MN, P54/58N, PMW1, RCC-associated antigen G250, RCC-associated protein G250, Renal cell carcinoma-associated antigen G250 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Carbonic Anhydrase IX/CA9 (NP_001207.2; Met 1-Asp 414). | |
0.1 mL | |
Cancer, HIF Target Genes, Hypoxia, Lipid and Metabolism, Metastasis, Renal Cell Carcinoma Cell Markers | |
768 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, ELISA | |
3 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction