Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carbonic Anhydrase IX/CA9 Antibody (CA9/2993R), DyLight 350, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP308407UV
Description
Carbonic Anhydrase IX/CA9 Monoclonal antibody specifically detects Carbonic Anhydrase IX/CA9 in Human, Equine samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| Carbonic Anhydrase IX/CA9 | |
| Monoclonal | |
| DyLight 350 | |
| CAIX, CA-IX, Carbonate dehydratase IX, carbonic anhydrase 9, carbonic anhydrase IX, carbonic dehydratase, EC 4.2.1.1, G250, Membrane antigen MN, MN, P54/58N, PMW1, RCC-associated antigen G250, RCC-associated protein G250, Renal cell carcinoma-associated antigen G250 | |
| Recombinant full-length human Carbonic Anhydrase IX/CA9 protein (Uniprot: Q16790) | |
| 100 μg | |
| Cancer, HIF Target Genes, Hypoxia, Lipid and Metabolism, Metastasis, Renal Cell Carcinoma Cell Markers | |
| 768 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| CA9/2993R | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Equine | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction