Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carbonic Anhydrase IX/CA9 Antibody (PN-15), Novus Biologicals™

Mouse Monoclonal Antibody
$268.00 - $524.00
Specifications
Antigen | Carbonic Anhydrase IX/CA9 |
---|---|
Clone | PN-15 |
Concentration | 0.2mg/mL |
Dilution | Western Blot 0.5 - 1.0 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, Immunofluorescence 1 - 2 ug/ml |
Applications | Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP24523800
![]() |
Novus Biologicals
NBP2452380.02MG |
0.02 mg |
Each for $268.00
|
|
|||||
NBP24523801
![]() |
Novus Biologicals
NBP2452380.1MG |
0.1 mg |
Each for $524.00
|
|
|||||
NBP24523802
![]() |
Novus Biologicals
NBP2452380.2MG |
0.2 mg | N/A | N/A | N/A | ||||
Description
Ensure accurate, reproducible results in Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence
Carbonic Anhydrase IX/CA9 Monoclonal specifically detects Carbonic Anhydrase IX/CA9 in Human, Equine samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunofluorescence.Specifications
Carbonic Anhydrase IX/CA9 | |
0.2mg/mL | |
Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence | |
Unconjugated | |
Mouse | |
Cancer, Cellular Markers, HIF Target Genes, Hypoxia, Lipid and Metabolism, Signal Transduction, Vision | |
10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
CA-IX, CAIXcarbonic anhydrase 9, Carbonate dehydratase IX, carbonic anhydrase IXpMW1, carbonic dehydratase, EC 4.2.1.1, G250, Membrane antigen MN, MNRCC-associated antigen G250, P54/58N, RCC-associated protein G250, Renal cell carcinoma-associated antigen G250 | |
CA9 | |
IgG2b κ | |
Protein A or G purified | |
Recognizes a glycoprotein of ∼200kDa, identified as carbonic anhydrase IX (CAIX/gp200). Its epitope resides in the carbohydrate domain of gp200. It shows no significant cross-reactivity with other carbohydrate determinants, such as the Lewis blood group antigens, epithelial membrane antigen, HMFG, and AB blood group antigens. In normal kidney, gp200 is localized along the brush border of the pars convoluta and pars recta segments of the proximal tubule, as well as focally along the luminal surface of Bowman's capsule adjoining the outgoing proximal tubule. Reportedly, gp200 is expressed by 93% of primary and 84% of metastatic renal cell carcinomas. This MAb may be useful in the investigations of carcinomas of proximal nephrogenic differentiation especially those showing tubular differentiation. |
PN-15 | |
Western Blot 0.5 - 1.0 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, Immunofluorescence 1 - 2 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human, Equine | |
Q16790 | |
768 | |
Microsomal fraction of human renal cortical tissue homogenate | |
Primary | |
Store at 4C. | |
55 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title