Learn More
Invitrogen™ Carbonic Anhydrase IX Polyclonal Antibody

Description
Immunogen sequence: PCAQGVIWTV FNQTVMLSAK QLHTLSDTLW GPGDSRLQLN FRATQPLNGR VIEASFPAGV DSSPRAAEPV QLNSCL Highest antigen sequence indentity to the following orthologs: Rat - 80%, Mouse - 80%.
Specifications
Specifications
| Antigen | Carbonic Anhydrase IX |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | CA9 |
| Gene Accession No. | Q16790 |
| Gene Alias | Ca9; CA-9; CAIX; CA-IX; Car9; Carbonate dehydratase IX; carbonic anhydrase 9; Carbonic anhydrase IX; carbonic dehydratase; G250; Membrane antigen MN; membrane antigen MN homolog; MN; MN/CA9; P54 / 58N; P54/58N; pMW1; RCC-associated antigen G250; RCC-associated protein G250; renal cell carcinoma-associated antigen G250 |
| Gene Symbols | CA9 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.