Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Carbonic Anhydrase VA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA580394
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA580394 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA580394 Supplier Invitrogen™ Supplier No. PA580394
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human Carbonic Anhydrase VA/CA5A.

Carbonic anhydrases (CAs) are members of a large family of zinc metalloenzymes that catalyze the reversible hydration of carbon dioxide. CAs are involved in a variety of biological processes, including respiration, calcification, acid-base balance, bone resorption, and the formation of aqueous humor, cerebrospinal fluid, saliva, and gastric juice. They show extensive diversity in distribution and in their subcellular localization. The human CA5A gene encodes for the CA V protein which is expressed in the mitochondrial matrix in liver tissues, but can also be detected in many other tissues. In ureagenesis, the CA V protein may be involved in providing bicarbonate to carbamoyl phosphate synthetase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Carbonic Anhydrase VA
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with no preservative
Gene CA5A
Gene Accession No. P35218
Gene Alias CA Y; CA5; CA5A; CA5AD; Car5; Car5a; Carbonate dehydratase VA; carbonic anhydrase 5 mitochondrial; carbonic anhydrase 5, mitochondrial; carbonic anhydrase 5A; carbonic anhydrase 5a, mitochondrial; carbonic anhydrase 5b, mitochondrial; carbonic anhydrase V, mitochondrial; carbonic anhydrase VA; carbonic anhydrase VA, mitochondrial; carbonic dehydratase; CAV; CAVA; CA-VA; GS1-21A4.1
Gene Symbols CA5A
Host Species Rabbit
Immunogen Recombinant Human Carbonic Anhydrase VA/CA5A protein (Ala40-Ser305).
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 763
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.