Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Carbonic Anhydrase VA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589996
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589996 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589996 Supplier Invitrogen™ Supplier No. PA589996
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CAWQTSNNTL HPLWTVPVSV PGGTRQSPIN IQWRDSVYDP QLKPLRVSYE AASCLYIWNT GYLFQVEFDD ATEASGISGG PLENHYRLKQ FHFHWGAVNE GGSEHTVDGH AYPAELHLVH WNSVKYQNYK EAVVGENGLA VIGVFLKLGA HHQTLQRLVD ILPEIKHKDA RAAMRPFDPS TLLPTCWDYW TYAGSLTTPP LTESVTWIIQ KEPVEVAPSQ LSAFRTLLFS ALGEEEKMMV NNYRPLQPLM NRKVWASFQA TNEGTRS; Positive Samples: K-562, LO2, HepG2; Cellular Location: Mitochondrion.

Carbonic anhydrases (CAs) are members of a large family of zinc metalloenzymes that catalyze the reversible hydration of carbon dioxide. CAs are involved in a variety of biological processes, including respiration, calcification, acid-base balance, bone resorption, and the formation of aqueous humor, cerebrospinal fluid, saliva, and gastric juice. They show extensive diversity in distribution and in their subcellular localization. The human CA5A gene encodes for the CA V protein which is expressed in the mitochondrial matrix in liver tissues, but can also be detected in many other tissues. In ureagenesis, the CA V protein may be involved in providing bicarbonate to carbamoyl phosphate synthetase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Carbonic Anhydrase VA
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene Car5a
Gene Accession No. P23589
Gene Alias CA Y; CA5; CA5A; CA5AD; Car5; Car5a; Carbonate dehydratase VA; carbonic anhydrase 5 mitochondrial; carbonic anhydrase 5, mitochondrial; carbonic anhydrase 5A; carbonic anhydrase 5a, mitochondrial; carbonic anhydrase 5b, mitochondrial; carbonic anhydrase V, mitochondrial; carbonic anhydrase VA; carbonic anhydrase VA, mitochondrial; carbonic dehydratase; CAV; CAVA; CA-VA; GS1-21A4.1
Gene Symbols Car5a
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 39-305 of human CA5A (NP_0017301).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12352
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.