Learn More
Invitrogen™ Carbonic Anhydrase VA Polyclonal Antibody, Invitrogen™
Description
Specifications
Specifications
| Antigen | Carbonic Anhydrase VA |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 |
| Gene | Car5a |
| Gene Accession No. | P23589, P43165 |
| Gene Alias | CA Y; CA5; CA5A; CA5AD; Car5; Car5a; Carbonate dehydratase VA; carbonic anhydrase 5 mitochondrial; carbonic anhydrase 5, mitochondrial; carbonic anhydrase 5A; carbonic anhydrase 5a, mitochondrial; carbonic anhydrase 5b, mitochondrial; carbonic anhydrase V, mitochondrial; carbonic anhydrase VA; carbonic anhydrase VA, mitochondrial; carbonic dehydratase; CAV; CAVA; CA-VA; GS1-21A4.1 |
| Gene Symbols | CA5A, Car5a |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.