Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carbonic Anhydrase VB/CA5B Antibody (008), Alexa Fluor™ 647, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290090AF647
Description
Carbonic Anhydrase VB/CA5B Monoclonal specifically detects Carbonic Anhydrase VB/CA5B in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Carbonic Anhydrase VB/CA5B | |
Monoclonal | |
Alexa Fluor 647 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Has cross-reactivity in ELISA with: Human CA2 Human CA3 Human CA5A No cross-reactivity in ELISA with: Human CA8 Human CA4 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
8 | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
Carbonate dehydratase VB, carbonic anhydrase 5B, mitochondrial, Carbonic anhydrase VB, carbonic anhydrase VB, mitochondrial, carbonic dehydratase, CA-VB, EC 4.2.1.1, MGC39962 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Carbonic Anhydrase VB/CA5B (Q9Y2D0; Cys34-Pro317). | |
0.1 mL | |
Lipid and Metabolism | |
11238 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction