Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carbonic Anhydrase VB/CA5B Antibody (008), PE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290090PE
Description
Carbonic Anhydrase VB/CA5B Monoclonal antibody specifically detects Carbonic Anhydrase VB/CA5B in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
Carbonic Anhydrase VB/CA5B | |
Monoclonal | |
PE | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Immunohistochemistry (Paraffin) | |
008 | |
Immunohistochemistry-Paraffin | |
Carbonate dehydratase VB, carbonic anhydrase 5B, mitochondrial, Carbonic anhydrase VB, carbonic anhydrase VB, mitochondrial, carbonic dehydratase, CA-VB, EC 4.2.1.1, MGC39962 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Carbonic Anhydrase VB/CA5B (Uniprot#: Q9Y2D0; Cys34-Pro317). | |
0.1 mL | |
Lipid and Metabolism | |
11238 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction