Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carbonic Anhydrase VB/CA5B Antibody, Alexa Fluor™ 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299972AF350
Description
Carbonic Anhydrase VB/CA5B Polyclonal specifically detects Carbonic Anhydrase VB/CA5B in Human samples. It is validated for Western Blot, ELISA.Specifications
Carbonic Anhydrase VB/CA5B | |
Polyclonal | |
Western Blot, ELISA | |
Carbonate dehydratase VB, carbonic anhydrase 5B, mitochondrial, Carbonic anhydrase VB, carbonic anhydrase VB, mitochondrial, carbonic dehydratase, CA-VB, EC 4.2.1.1, MGC39962 | |
Produced in rabbits immunized with purified, recombinant Carbonic Anhydrase VB/CA5B (Q9Y2D0; Cys34-Pro317). | |
0.1 mL | |
Lipid and Metabolism | |
11238 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, ELISA | |
Alexa Fluor 350 | |
50mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction