Learn More
Carbonic Anhydrase VI Monoclonal Antibody (401809)

Description
Carbonic Anhydrase VI Monoclonal Antibody for Flow
Specifications
Specifications
| Antigen | Carbonic Anhydrase VI |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 401809 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | CA6 |
| Gene Accession No. | P23280 |
| Gene Alias | CA6; CA-6; Car6; Carbonate dehydratase VI; carbonic anhydrase 6; Carbonic anhydrase VI; carbonic anhydrase VI nirs variant 2; Carbonic anhydrase6; CA-VI; DOC1; GUSTIN; salivary carbonic anhydrase; Secreted carbonic anhydrase |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.