Learn More
Invitrogen™ Carbonic Anhydrase VI Polyclonal Antibody

Description
Carbonic Anhydrase VI Polyclonal Antibody for Western Blot, IHC (P), Flow
Specifications
Specifications
| Antigen | Carbonic Anhydrase VI |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | CA6 |
| Gene Accession No. | P23280 |
| Gene Alias | CA6; CA-6; Car6; Carbonate dehydratase VI; carbonic anhydrase 6; Carbonic anhydrase VI; carbonic anhydrase VI nirs variant 2; Carbonic anhydrase6; CA-VI; DOC1; GUSTIN; salivary carbonic anhydrase; Secreted carbonic anhydrase |
| Gene Symbols | CA6 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.