Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Carbonic Anhydrase VI Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583027
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583027 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583027 Supplier Invitrogen™ Supplier No. PA583027
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SEISGSEHTV DGIRHVIEIH IVHYNSKYKS YDIAQDAPDG LAVLAAFVEV KNYPENTYYS NFISHLANIK YPGQRTTLTG LDVQDMLPRN LQHYYTY Highest antigen sequence indentity to the following orthologs: Rat - 62%, Mouse - 52%.

The protein encoded by this gene is one of several isozymes of carbonic anhydrase. This protein is found only in salivary glands and saliva and protein may play a role in the reversible hydratation of carbon dioxide though its function in saliva is unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Carbonic Anhydrase VI
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CA6
Gene Accession No. P23280
Gene Alias CA6; CA-6; Car6; Carbonate dehydratase VI; carbonic anhydrase 6; Carbonic anhydrase VI; carbonic anhydrase VI nirs variant 2; Carbonic anhydrase6; CA-VI; DOC1; GUSTIN; salivary carbonic anhydrase; Secreted carbonic anhydrase
Gene Symbols CA6
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CA6. Recombinant protein control fragment (Product #RP-94191).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 765
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.