Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carbonic Anhydrase VIII/CA8 Antibody (001), DyLight 650, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290709C
Description
Carbonic Anhydrase VIII/CA8 Monoclonal specifically detects Carbonic Anhydrase VIII/CA8 in Mouse samples. It is validated for ELISA, Sandwich ELISA Detection.Specifications
Carbonic Anhydrase VIII/CA8 | |
Monoclonal | |
DyLight 650 | |
CALScarbonate dehydratase, CAMRQ3, carbonic anhydrase VIIIMGC99509, carbonic anhydrase-like sequence, carbonic anhydrase-related protein, CARPCA-related protein, CA-VIII, MGC120502 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Carbonic Anhydrase VIII/CA8 (P28651; Met1-Gln291). | |
0.1 mL | |
Lipid and Metabolism | |
767 | |
Mouse | |
Purified |
ELISA | |
1 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Has cross-reactivity in ELISA with: Human CA8 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction