Learn More
Carbonic Anhydrase VIII/CA8 Antibody, DyLight 405, Novus Biologicals™
SDP
Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals NBP298350V
Catalog No. NB104072
Only null left
Add to Cart
Description
Description
Carbonic Anhydrase VIII/CA8 Polyclonal specifically detects Carbonic Anhydrase VIII/CA8 in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
Carbonic Anhydrase VIII/CA8
Polyclonal
50 mM Sodium Borate
Rabbit
Affinity purified
RUO
Primary
Mouse
Purified
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
DyLight 405
CALScarbonate dehydratase, CAMRQ3, carbonic anhydrase VIIIMGC99509, carbonic anhydrase-like sequence, carbonic anhydrase-related protein, CARPCA-related protein, CA-VIII, MGC120502
Produced in rabbits immunized with purified, recombinant Carbonic Anhydrase VIII/CA8 (P28651; Met1-Gln291).
0.1 mL
Lipid and Metabolism
767
Store at 4°C in the dark.
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.