Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carbonic Anhydrase VIII/CA8 Antibody, DyLight 650, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298350C
Description
Carbonic Anhydrase VIII/CA8 Polyclonal specifically detects Carbonic Anhydrase VIII/CA8 in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Carbonic Anhydrase VIII/CA8 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 650 | |
CALScarbonate dehydratase, CAMRQ3, carbonic anhydrase VIIIMGC99509, carbonic anhydrase-like sequence, carbonic anhydrase-related protein, CARPCA-related protein, CA-VIII, MGC120502 | |
Produced in rabbits immunized with purified, recombinant Carbonic Anhydrase VIII/CA8 (P28651; Met1-Gln291). | |
0.1 mL | |
Lipid and Metabolism | |
767 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction