Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carbonic Anhydrase VIII Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547267
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547267 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547267 Supplier Thermo Scientific Supplier No. PA547267
Add to Cart
Add to Cart

Carbonic Anhydrase VIII Polyclonal Antibody

Carbonic Anhydrase VIII Polyclonal Antibody for Western Blot, IP

TRUSTED_SUSTAINABILITY

Specifications

Antigen Carbonic Anhydrase VIII
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene Ca8
Gene Accession No. P28651, P35219
Gene Alias AW546993; Ca8; Cals; Cals1; CAMRQ3; Car8; carbonate dehydratase; carbonic anhydrase 8; carbonic anhydrase VIII; carbonic anhydrase-like sequence; carbonic anhydrase-like sequence 1; carbonic anhydrase-related protein; CA-related protein; CARP; CA-RP; CARP VIII; CA-RP VIII; CA-VIII; hypothetical protein LOC550233; LOW QUALITY PROTEIN: carbonic anhydrase-related protein; MGC120502; MGC99509; wdl; zgc:110118
Gene Symbols Ca8, Car8
Host Species Goat
Immunogen E. coli-derived recombinant human Carbonic AnhydraseVIII/CA8 Ala2-Gln290.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12319, 767
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.