Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carbonic Anhydrase X/CA10 Antibody (7H8F12E6), Alexa Fluor™ 532 , Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP306193AF532
Description
Carbonic Anhydrase X/CA10 Monoclonal antibody specifically detects Carbonic Anhydrase X/CA10 in Human samples. It is validated for ELISASpecifications
Carbonic Anhydrase X/CA10 | |
Monoclonal | |
Alexa Fluor 532 | |
carbonic anhydrase X, carbonic anhydrase-related protein 10, Carbonic anhydrase-related protein X, CARP X, CA-RPX, CARPXCA-RP X, Cerebral protein 15, cerebral protein-15, HUCEP-15 | |
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Carbonic Anhydrase X/CA10 (NP_001076002.1; Met 1-Asn 300). | |
0.1 mL | |
Primary | |
Human | |
Purified |
ELISA | |
7H8F12E6 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
56934 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction