Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carbonic Anhydrase X/CA10 Antibody, DyLight 650, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298120C
Description
Carbonic Anhydrase X/CA10 Polyclonal specifically detects Carbonic Anhydrase X/CA10 in Mouse samples. It is validated for Western Blot, ELISA.Specifications
Carbonic Anhydrase X/CA10 | |
Polyclonal | |
Western Blot, ELISA | |
carbonic anhydrase X, carbonic anhydrase-related protein 10, Carbonic anhydrase-related protein X, CARP X, CA-RPX, CARPXCA-RP X, Cerebral protein 15, cerebral protein-15, HUCEP-15 | |
Produced in rabbits immunized with purified, recombinant Carbonic Anhydrase X/CA10 (P61215; Met1-Asn300). | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
Western Blot, ELISA | |
DyLight 650 | |
50mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
56934 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction