Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Carbonic Anhydrase X Monoclonal Antibody (7H8F12E6), Invitrogen™

Catalog No. PIMA543507
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA543507 50 μL
PIMA542996 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA543507 Supplier Invitrogen™ Supplier No. MA543507
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Specificity: Human CA10/CARPX

This gene encodes a protein that belongs to the carbonic anhydrase family of zinc metalloenzymes, which catalyze the reversible hydration of carbon dioxide in various biological processes. The protein encoded by this gene is an acatalytic member of the alpha-carbonic anhydrase subgroup, and it is thought to play a role in the central nervous system, especially in brain development. Multiple transcript variants encoding the same protein have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Carbonic Anhydrase X
Applications ELISA
Classification Monoclonal
Clone 7H8F12E6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene CA10
Gene Accession No. Q9NS85
Gene Alias 2700029L05Rik; BB085816; Ca10; Car10; carbonic anhydrase 10; carbonic anhydrase X; carbonic anhydrase-related protein 10; Carbonic anhydrase-related protein X; CARP X; CA-RP X; CARPX; CA-RPX; Cerebral protein 15; cerebral protein-15; HUCEP-15; UNQ533/PRO1076
Gene Symbols CA10
Host Species Mouse
Immunogen Recombinant Human CA10 protein.
Purification Method Protein A
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56934
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.