Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carbonic Anhydrase X Polyclonal Antibody
Carbonic Anhydrase X Polyclonal Antibody
Supplier: Thermo Scientific PA566106
Description
Carbonic Anhydrase X Polyclonal Antibody for ICC/IF

Specifications
Carbonic Anhydrase X | |
Polyclonal | |
Unconjugated | |
CA10 | |
2700029L05Rik; BB085816; Ca10; Car10; carbonic anhydrase 10; carbonic anhydrase X; carbonic anhydrase-related protein 10; Carbonic anhydrase-related protein X; CARP X; CA-RP X; CARPX; CA-RPX; Cerebral protein 15; cerebral protein-15; HUCEP-15; UNQ533/PRO1076 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
56934 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NS85 | |
CA10 | |
Recombinant Human Carbonic Anhydrase X. Recombinant protein control fragment (Product #RP-106797). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction