Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carbonic Anhydrase XII/CA12 Antibody (001), HRP , Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290350H
Description
Carbonic Anhydrase XII/CA12 Monoclonal antibody specifically detects Carbonic Anhydrase XII/CA12 in Mouse samples. It is validated for ELISASpecifications
Carbonic Anhydrase XII/CA12 | |
Monoclonal | |
HRP | |
Carbonate dehydratase XII, carbonic anhydrase 12, carbonic anhydrase XIICAXII, carbonic dehydratase, CA-XII, EC 4.2.1.1, FLJ20151, HsT18816, Tumor antigen HOM-RCC-3.1.3 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Carbonic Anhydrase XII/CA12 (NP_848483.2; Met1-Ser301). | |
0.1 mL | |
Lipid and Metabolism | |
771 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
001 | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction