Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carbonic Anhydrase XII/CA12 Antibody (012), Janelia Fluor™ 549, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290352JF549
Description
Carbonic Anhydrase XII/CA12 Monoclonal specifically detects Carbonic Anhydrase XII/CA12 in Mouse samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Carbonic Anhydrase XII/CA12 | |
Monoclonal | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
12 | |
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
Carbonate dehydratase XII, carbonic anhydrase 12, carbonic anhydrase XIICAXII, carbonic dehydratase, CA-XII, EC 4.2.1.1, FLJ20151, HsT18816, Tumor antigen HOM-RCC-3.1.3 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Carbonic Anhydrase XII/CA12 (NP_848483.2; Met 1-Ser 301). | |
0.1 mL | |
Lipid and Metabolism | |
771 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction