Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Carbonic Anhydrase XII/CA12 Antibody Pair [HRP]
SDP

Catalog No. NB010037 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
15 Plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB010037 15 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB010037 Supplier Novus Biologicals™ Supplier No. NBP27949915PLATES

Mouse Monoclonal Antibody Pair

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human Carbonic Anhydrase XII/CA12.

Specifications

Assay Range 7.8 pg/mL
Assay Sensitivity 7.81-500 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human Carbonic Anhydrase XII/CA12.
Target Species Human
For Use With (Application) Sandwich ELISA
Gene Alias Carbonate dehydratase XII, carbonic anhydrase 12, carbonic anhydrase XIICAXII, carbonic dehydratase, CA-XII, EC 4.2.1.1, FLJ20151, HsT18816, Tumor antigen HOM-RCC-3.1.3
Gene ID (Entrez) 771
Gene Symbol CA12
Host Species Mouse
Kit Contents
  • Detection Antibody: 0.25 mg/mL mouse anti-Carbonic Anhydrase XII/CA12 monoclonal antibody conjugated to horseradish-peroxidase (HRP).
  • Capture Antibody: 90 μg/mL of mouse anti-Carbonic Anhydrase XII/CA12 monoclonal antibody,
  • Standard: Each vial contains 37 ng of recombinant Carbonic Anhydrase XII/CA12.
  • Quantity 15 Plates
    Regulatory Status RUO
    Research Discipline Lipid and Metabolism
    Storage Requirements Storage is content dependent.
    Target Carbonic Anhydrase XII/CA12
    Technique 1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in CBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve usi ng 2-fold serial dilutions in sample dilution buffer, and a high standard of 0.5 ng/mL is recommended.
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.