Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Carbonic Anhydrase XII Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587698
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587698 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587698 Supplier Invitrogen™ Supplier No. PA587698
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: APVNGSKWTY FGPDGENSWS KKYPSCGGLL QSPIDLHSDI LQYDASLTPL EFQGYNLSAN KQFLLTNNGH SVKLNLPSDM HIQGLQSRYS ATQLHLHWGN PNDPHGSEHT VSGQHFAAEL HIVHYNSDLY PDASTASNKS EGLAVLAVLI EMGSFNPSYD KIFSHLQHVK YKGQEAFVPG FNIEELLPER TAEYYRYRGS LTTPPCNPTV LWTVFRNPVQ ISQEQLLALE TALYCTHMDD PSPREMINNF RQVQKFDERL VYTSFSQVQV CTAAGLS; Positive Samples: HT-29; Cellular Location: Membrane, Single-pass type I membrane protein.

Carbonic Anhydrase XII is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Carbonic Anhydrase XII
Applications Western Blot
Classification Polyclonal
Concentration 1.84 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CA12
Gene Accession No. O43570, Q8CI85
Gene Alias 2310047E01Rik; AI314958; CA12; Car12; Carbonate dehydratase XII; carbonic anhydrase 12; Carbonic anhydrase XII; carbonic anyhydrase 12; carbonic dehydratase; CAXII; CA-XII; HsT18816; membrane-bound carbonic anhydrase 12; T18816; tumor antigen HOM-RCC-3.1.3
Gene Symbols CA12, Car12
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 25-301 of human CA12 (NP_0012091).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 363085, 76459, 771
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.