Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carbonic Anhydrase XIII/CA13 Antibody (103), mFluor Violet 450 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289511MFV450
Description
Carbonic Anhydrase XIII/CA13 Monoclonal antibody specifically detects Carbonic Anhydrase XIII/CA13 in Human samples. It is validated for ELISASpecifications
Carbonic Anhydrase XIII/CA13 | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
103 | |
ELISA | |
carbonic anhydrase 13, carbonic anhydrase XIIICarbonate dehydratase XIII, CAXIII, CA-XIII, EC 4.2.1.1, FLJ37995, MGC59868 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Carbonic Anhydrase XIII/CA13 (Accession#: NP_940986.1; Met1-His262). | |
0.1 mL | |
Lipid and Metabolism | |
377677 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction