Learn More
Invitrogen™ Carbonic Anhydrase XIII Recombinant Rabbit Monoclonal Antibody (103), Invitrogen™

Description
Specificity: Human Carbonic Anhydrase XIII/CA13
Specifications
Specifications
| Antigen | Carbonic Anhydrase XIII |
| Applications | ELISA |
| Classification | Recombinant Monoclonal |
| Clone | 103 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative |
| Gene | Ca13 |
| Gene Accession No. | Q8N1Q1 |
| Gene Alias | 2310075C21Rik; Ca13; Car13; Carbonate dehydratase XIII; carbonic anhydrase 13; Carbonic anhydrase XIII; CAXIII; CA-XIII; RGD1560453 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.