Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carbonic Anhydrase XIV/CA14 Antibody (033), DyLight 350 , Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290349UV
Description
Carbonic Anhydrase XIV/CA14 Monoclonal antibody specifically detects Carbonic Anhydrase XIV/CA14 in Mouse samples. It is validated for ELISASpecifications
Carbonic Anhydrase XIV/CA14 | |
Monoclonal | |
DyLight 350 | |
Carbonate dehydratase XIV, carbonic anhydrase XIVcarbonic anhydrase 14, carbonic dehydratase, CAXiV, CA-XIV, EC 4.2.1.1 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Carbonic Anhydrase XIV/CA14 (NP_035927.1; Met 1-Met 290). | |
0.1 mL | |
Lipid and Metabolism | |
23632 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
033 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction