Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carbonic Anhydrase XIV/CA14 Antibody (110), Alexa Fluor™ 532, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289509AF532
Description
Carbonic Anhydrase XIV/CA14 Monoclonal antibody specifically detects Carbonic Anhydrase XIV/CA14 in Human samples. It is validated for ELISA, ELISASpecifications
Carbonic Anhydrase XIV/CA14 | |
Monoclonal | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
110 | |
ELISA, Sandwich ELISA | |
Carbonate dehydratase XIV, carbonic anhydrase XIVcarbonic anhydrase 14, carbonic dehydratase, CAXiV, CA-XIV, EC 4.2.1.1 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Carbonic Anhydrase XIV/CA14 (NP_036245.1; Met1-Met290). | |
0.1 mL | |
Lipid and Metabolism | |
23632 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction