Learn More
Invitrogen™ Carbonic Anhydrase XIV Monoclonal Antibody (312908)

Description
In direct ELISAs and Western blots, approximately 50% cross-reactivity with recombinant mouse CA14, 10% cross-reactivity with recombinant human (rh) CA5A, and no cross-reactivity with rhCA1, 2, 3, 4, 6, 7, 8, 9, 10, 12, or 13 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.
Specifications
Specifications
| Antigen | Carbonic Anhydrase XIV |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | 312908 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | CA14 |
| Gene Accession No. | Q9ULX7 |
| Gene Alias | AW536446; CA XIV; CA14; Car14; Carbonate dehydratase XIV; carbonic anhydrase 14; Carbonic anhydrase XIV; carbonic dehydratase; Catm; CAXiV; CA-XIV; UNQ690/PRO1335 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.