Learn More
Invitrogen™ Carbonic Anhydrase XIV Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA547336
Description
In direct ELISAs and Western blots, approximately 30% cross-reactivity with recombinant human (rh) CA14 is observed and less than 5% cross-reactivity with rhCA1, -2, -3, -6, -8, -10, -13, recombinant mouse CA4, -9, and -12 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
CA14 belongs a large family of zinc metalloenzymes that catalyze the versible hydration of carbon dioxide. The protein is predicted to be a type I membrane protein and shares highest sequence similarity with the other transmembrane CA isoform, CA XII; however, they have different patterns of tissue-specific expression and thus may play different physiologic roles.
Specifications
Carbonic Anhydrase XIV | |
Polyclonal | |
Unconjugated | |
CA14 | |
AW536446; CA XIV; CA14; Car14; Carbonate dehydratase XIV; carbonic anhydrase 14; Carbonic anhydrase XIV; carbonic dehydratase; Catm; CAXiV; CA-XIV; UNQ690/PRO1335 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
23632, 23831, 791259 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunoprecipitation, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q9ULX7, Q9WVT6 | |
CA14, Car14 | |
Mouse myeloma cell line NS0-derived recombinant mouse Carbonic AnhydraseXIV/CA14 Ala16-Met290. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.