Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Carbonic Anhydrase XIV Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101490
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101490 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101490 Supplier Invitrogen™ Supplier No. PA5101490
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CA14.

CA14 belongs a large family of zinc metalloenzymes that catalyze the versible hydration of carbon dioxide. The protein is predicted to be a type I membrane protein and shares highest sequence similarity with the other transmembrane CA isoform, CA XII; however, they have different patterns of tissue-specific expression and thus may play different physiologic roles.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Carbonic Anhydrase XIV
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CA14
Gene Accession No. Q9ULX7, Q9WVT6
Gene Alias AW536446; CA XIV; CA14; Car14; Carbonate dehydratase XIV; carbonic anhydrase 14; Carbonic anhydrase XIV; carbonic dehydratase; Catm; CAXiV; CA-XIV; UNQ690/PRO1335
Gene Symbols CA14, Car14
Host Species Rabbit
Immunogen A synthesized peptide derived from human CA14(Accession Q9ULX7), corresponding to amino acid residues G161-R211.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23632, 23831, 791259
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.