Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carbonylchlorobis(triphenylphosphine)iridium(I)

Catalog No. AA3941303
Change view
Click to view available options
Quantity:
0.25 g
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3941303 1 g
AA3941301 0.25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3941303 Supplier Thermo Scientific Chemicals Supplier No. 03941303
Add to Cart

CAS: 14871-41-1 | C37H30ClIrOP2 | 780.26 g/mol

Carbonylchlorobis(triphenylphosphine)iridium(I) is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14871-41-1
Molecular Formula C37H30ClIrOP2
Molecular Weight (g/mol) 780.26
MDL Number MFCD00009590
InChI Key PEPSDKMVHYXIMJ-UHFFFAOYSA-M
Synonym carbonylchlorobis triphenylphosphine iridium i
PubChem CID 78076234
IUPAC Name iridium;methanone;triphenylphosphane;chloride
SMILES Cl[Ir+].[C-]#[O+].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 215°C (decomposition)
Quantity 1 g
UN Number UN2811
Solubility Information Soluble in chloroform, toluene; Slightly soluble in acetone, alcohol
Formula Weight 780.27
Physical Form Crystalline
Chemical Name or Material Carbonylchlorobis(triphenylphosphine)iridium(I)
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 238-941-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.