Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carbonyldihydridotris(triphenylphosphine)ruthenium(II), 99%

Catalog No. AA4052409
Change view
Click to view available options
Quantity:
0.5 g
2 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4052409 10 g
AA4052402 0.5 g
AA4052404 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4052409 Supplier Thermo Scientific Chemicals Supplier No. 4052409

CAS: 25360-32-1 | C55H47OP3Ru | 917.97 g/mol

Carbonyldihydridotris(triphenylphosphine)ruthenium(II) is used as Catalyst for conversion of 1,4-alkynediols into pyrroles, Catalyst for oxidation of primary alcohols to methyl esters, Catalyst for rearrangement of oximes to amides, Catalyst for C-C coupling via directed C-H activation, Catalyst for enyne cyclization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25360-32-1
Molecular Formula C55H47OP3Ru
Molecular Weight (g/mol) 917.97
MDL Number MFCD00015870 MFCD00015870
InChI Key MLIYPCQSOXNTLJ-UHFFFAOYSA-N
PubChem CID 53384314
IUPAC Name carbon monoxide;hydride;ruthenium(2+);triphenylphosphane
SMILES [RuH2].[C-]#[O+].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 161°C to 163°C
Color White
Assay Percent Range 99%
Quantity 10 g
Sensitivity Light and moisture sensitive
Solubility Information Insoluble in water.
Formula Weight 917.97
Physical Form Crystals
Chemical Name or Material Carbonyldihydridotris(triphenylphosphine)ruthenium(II)
Hazard Category H302+H312+H332
Hazard Statement In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing and gloves.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.