Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Carbopack™ B Thermal Desorption Tubes

Catalog No. 111005374 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Material:
Glass
Stainless Steel
Description:
Pre-conditioned
Pre-conditioned/Sealed with Brass Endcaps
Unconditioned
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Description
11-100-5374 Stainless Steel Unconditioned
11-100-5367 Glass Pre-conditioned
11-100-5352 Glass Pre-conditioned/Sealed with Brass Endcaps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-100-5374 Supplier MilliporeSigma™ Supelco™ Supplier No. 30135U
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Use for the trapping and extraction of herbicides, simazine, and atrazine in water samples followed by high-performance liquid chromatography (HPLC) analysis

  • Surface Area: 100 m2/g
  • USP Code: S12
TRUSTED_SUSTAINABILITY

Specifications

Includes CarbopackTM B Stainless Steel TD Tubes (10)
Overall Length (Metric) 8.89 cm
Diameter (Metric) Outer 6.35 mm
For Use With (Equipment) PerkinElmer, Markes, DANI, OI Analytical, and Shimadzu Instruments
Material Stainless Steel
For Use With (Application) Air Monitoring, Sampling a Wide Range of VOCs
Matrix Carbopack™ Graphitized Carbon Black (GCB)
Overall Length (English) 3 1/2 in.
Product Line Carbopack
Type Thermal Sorption Tube
Packaging Quantity 10/Pk.
Diameter (English) Outer 1/4 in.
Description Unconditioned
Show More Show Less

Carbopack is a trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.