Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Carbopack X™ Thermal Desorption Tubes

Catalog No. 111005362 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Description:
Pre-conditioned
Pre-conditioned/Sealed with Brass Endcaps
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
11-100-5362 Pre-conditioned
11-100-5354 Pre-conditioned/Sealed with Brass Endcaps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-100-5362 Supplier MilliporeSigma™ Supelco™ Supplier No. 29537U
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Suitable for use with Air Monitoring

  • For analyte group C3-C5
TRUSTED_SUSTAINABILITY

Specifications

Overall Length (Metric) 88.9 mm
Includes 10 Carbopack™ X Thermal Desorption Tubes
Diameter (Metric) Outer 6.35 mm
Overall Length (English) 3.5 in.
For Use With (Equipment) PerkinElmer, Markes, DANI, OI Analytical, and Shimadzu Instruments
Material Glass
Product Line Carbopack
Matrix Carbopack™ x Graphitized Carbon Black (GCB)
Diameter (English) Outer 1/4 in.
Packaging Quantity 10/Pk.
For Use With (Application) Air Monitoring
Methods EPA TO-17
Description Pre-conditioned
Type Thermal Desorption Tube
Show More Show Less

Carbopack is a trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.