Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Carborundum
SCI_ED

Catalog No. p-4416737 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
250 g
Packaging:
Solid
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25235 100 g Solid
S25235A 250 g Solid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S25235
Polishing compound

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 409-21-2
Molecular Formula CSi
Molecular Weight (g/mol) 40.10
MDL Number MFCD00049531
InChI Key HBMJWWWQQXIZIP-UHFFFAOYSA-N
Synonym Silicon Carbide
PubChem CID 9863
ChEBI CHEBI:29390
IUPAC Name methanidylidynesilylium
SMILES [C-]#[Si+]

Specifications

Quantity 100 g
Density 3.2 g/mL
Color Black to Colorless
Packaging Solid
Assay Percent Range 100% w/v
Formula Weight 40.09
Chemical Name or Material Carborundum
Safety handling Check Alert storage code Red
ShelfLife 5 Years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.